Learn More
Perkin Elmer US LLC Self-Aligning RF Load Coil Assembly for NexION 300/350 & ELAN 6x00/9000/DRCs
Catalog No. 509046860
Encompass_Preferred
This kit includes: 46 cm Copper Refrigeration Tube, Qty. 1 (W1038883) Brass Nut for 3.175 mm Swagelok Tube Fitting, Qty. 2 (WE011094) Brass Back Ferrule for 3.175 mm Swagelok Tube Fitting, Qty. 2 (WE011095) Brass Front Ferrule for 3.175 mm Swagelok Tube Fitting, Qty. 2 (WE011096) Coil Forming Installation Tool, Qty. 1 (WE014808) Nylon 14.2 cm Cable Tie, Qty. 2 (WE050011)
Catalog No. 50-904-6860 Supplier Perkin Elmer US LLC Supplier No. WE021816
This kit includes: 46 cm Copper Refrigeration Tube, Qty. 1 (W1038883) Brass Nut for 3.175 mm Swagelok Tube Fitting, Qty. 2 (WE011094) Brass Back Ferrule for 3.175 mm Swagelok Tube Fitting, Qty. 2 (WE011095) Brass Front Ferrule for 3.175 mm Swagelok Tube Fitting, Qty. 2 (WE011096) Coil Forming Installation Tool, Qty. 1 (WE014808) Nylon 14.2 cm Cable Tie, Qty. 2 (WE050011)