Learn More
Zymo Research Corporation Load N Go Quick-DNA/RNA HT Kit
SDP
Catalog No. 502865903
Encompass_Preferred
The Load N' Go Quick-DNA/RNA HT kit offers a streamlined solution for high-throughput nucleic acid extraction from a variety of biological and clinical samples, such as swabs, biological liquids, and tissues. Compatible with robotic sample processors, this kit ensures the efficient isolation of high-quality DNA and RNA, making it suitable for sensitive downstream applications, including Next-Gen sequencing and RT/qPCR, ultimately enhancing the precision and reliability of scientific research.
Catalog No. 50-286-5903 Supplier Zymo Research Corporation Supplier No. R2152
The Load N' Go Quick-DNA/RNA HT kit offers a streamlined solution for high-throughput nucleic acid extraction from a variety of biological and clinical samples, such as swabs, biological liquids, and tissues. Compatible with robotic sample processors, this kit ensures the efficient isolation of high-quality DNA and RNA, making it suitable for sensitive downstream applications, including Next-Gen sequencing and RT/qPCR, ultimately enhancing the precision and reliability of scientific research.