Learn More
Bioworld Loading Buffer (2X) Protein Gel, 5 mL
Catalog No. 50198706
Encompass_Preferred
An electrophoretic dye for denaturation of proteins and monitoring the front of running gel. Ultra Pure Grade Contents: 4% SDS and 20% glycerol. To make reducing add 200mM β-mercaptoethanol or DTT.DNase: None detected RNase: None detected Proteases: None detected Stability (200°C) >1 Year Storage: RT
Catalog No. 50-198-706 Supplier Bioworld Supplier No. 214700031
An electrophoretic dye for denaturation of proteins and monitoring the front of running gel. Ultra Pure Grade Contents: 4% SDS and 20% glycerol. To make reducing add 200mM β-mercaptoethanol or DTT.DNase: None detected RNase: None detected Proteases: None detected Stability (200°C) >1 Year Storage: RT