Learn More
Bioworld Loading Buffer (5x), 30% Glycerol, 5 mL
Catalog No. 50198708
Encompass_Preferred
For agarose gels. Excellent electroohoretic dye for most DNA and RNA application. Ultra Pure Grade Contains: 30% Glycerol DNase: None Detected RNase: None Detected Stability (+4°C) >1 year Storage: 2-8°C
Catalog No. 50-198-708 Supplier Bioworld Supplier No. 214700042
For agarose gels. Excellent electroohoretic dye for most DNA and RNA application. Ultra Pure Grade Contains: 30% Glycerol DNase: None Detected RNase: None Detected Stability (+4°C) >1 year Storage: 2-8°C