Learn More
Bioworld Loading Buffer (5x), 30% Glycerol, 1 mL
Catalog No. 50198707
Encompass_Preferred
For agarose gels. Excellent electroohoretic dye for most DNA and RNA application. Ultra Pure Grade Contains: 30% Glycerol DNase: None Detected RNase: None Detected Stability (+4°C) >1 year Storage: 2-8°C
Catalog No. 50-198-707 Supplier Bioworld Supplier No. 214700041
For agarose gels. Excellent electroohoretic dye for most DNA and RNA application. Ultra Pure Grade Contains: 30% Glycerol DNase: None Detected RNase: None Detected Stability (+4°C) >1 year Storage: 2-8°C