Learn More
AdipoGen Loading Buffer 5X
SDP
Catalog No. 502563400
Encompass_Preferred
Chemical. Synthetic. Loading buffer for separation and visualization proteins with SDS-PAGE and Western Blot analysis.
Catalog No. 50-256-3400 Supplier AdipoGen Supplier No. AG10T0020L001
Chemical. Synthetic. Loading buffer for separation and visualization proteins with SDS-PAGE and Western Blot analysis.