Learn More
Bioworld Loading Buffer (5x), 40% Sucrose, 5 mL
Catalog No. 50198710
Encompass_Preferred
For agarose gels. Excellent electroohoretic dye for most DNA and RNA application. Ultra Pure Grade Contains 40% Sucrose DNase: None Detected RNase: None Detected Stability (+4°C) >1 year Storage: 2-8°C
Catalog No. 50-198-710 Supplier Bioworld Supplier No. 214700052
For agarose gels. Excellent electroohoretic dye for most DNA and RNA application. Ultra Pure Grade Contains 40% Sucrose DNase: None Detected RNase: None Detected Stability (+4°C) >1 year Storage: 2-8°C