Learn More
As One International Inc Loading Buffer Blue, 5 x 1 mL
Catalog No. 501656029
Encompass_Preferred
5x DNA loading buffer used for loading DNA samples onto agarose or polyacrylamide gels and tracking migration during electrophoresis. Blue tracking dye migrates at 100-300 bp
Catalog No. 50-165-6029 Supplier As One International Inc Supplier No. AO608204
5x DNA loading buffer used for loading DNA samples onto agarose or polyacrylamide gels and tracking migration during electrophoresis. Blue tracking dye migrates at 100-300 bp