Learn More
As One International Inc Loading Buffer Orange, 5 x 1 mL
Catalog No. 501656030
Encompass_Preferred
5x DNA loading buffer used for loading DNA samples onto agarose or polyacrylamide gels and tracking migration during electrophoresis. Orange tracking dye migrates at 50-80 bp
Catalog No. 50-165-6030 Supplier As One International Inc Supplier No. AO608304
5x DNA loading buffer used for loading DNA samples onto agarose or polyacrylamide gels and tracking migration during electrophoresis. Orange tracking dye migrates at 50-80 bp