Learn More
Bioworld Loading Dye Base, 5 or More, 5 mL
Catalog No. 50198713
Encompass_Preferred
Ultra Pure Grade Contains 50x Loading Dye Blend in H2O Mix with buffer and density agents before use. DNase: None Detected RNase: None Detected Stability (+20C): > 1year Storage: RT
Catalog No. 50-198-713 Supplier Bioworld Supplier No. 214700071
Ultra Pure Grade Contains 50x Loading Dye Blend in H2O Mix with buffer and density agents before use. DNase: None Detected RNase: None Detected Stability (+20C): > 1year Storage: RT