Learn More
Bioworld Loading Dye Base, 100 mL
Catalog No. 50198711
Encompass_Preferred
Ultra Pure Grade.Contains 50x Loading Dye Blend in buffer.Mix buffer, density agents, and SDS,MetOH before use.DNase: None Detected. RNase: None Detected. Stability (+20C): > 1year Storage: RTDyes only in Tris pH 8.5, without glycerol or sucrose.
Catalog No. 50-198-711 Supplier Bioworld Supplier No. 214700061
Ultra Pure Grade.Contains 50x Loading Dye Blend in buffer.Mix buffer, density agents, and SDS,MetOH before use.DNase: None Detected. RNase: None Detected. Stability (+20C): > 1year Storage: RTDyes only in Tris pH 8.5, without glycerol or sucrose.