Learn More
Kuhner Shaker Inc LOADS UP TO 25KG 25KG POSSIB
Catalog No. 502569928
Encompass
ISF1-ZC Shaker with humidity, cooling and CO2Temperature range: ambient -15C to 60CControl of 0.1C, Accuracy +/- 0.3C at 37CCO2 Control 0-20%, Increments of 0.1%, Accuracy +/- 0.42% @ 20% CO2Bi-directional humidity control to 90%RH, accuracy +/-1%User Selectable Orbit (12.5mm, 25mm, 50mm)Loads up to 25kg (>25kg possible if required)DC direct drive with single eccentric (no belt)100% stainless steel interiorDimensions: H771 x W1275 x D845mmWeight: 210kgFIVE YEAR WARRANTY HARDWARE1 YEAR WARRANTY ON SENSORS
Catalog No. 50-256-9928 Supplier Kuhner Shaker Inc Supplier No. SMZ1503C
Add to Cart
Add to Cart
Request a Quote
ISF1-ZC Shaker with humidity, cooling and CO2Temperature range: ambient -15C to 60CControl of 0.1C, Accuracy +/- 0.3C at 37CCO2 Control 0-20%, Increments of 0.1%, Accuracy +/- 0.42% @ 20% CO2Bi-directional humidity control to 90%RH, accuracy +/-1%User Selectable Orbit (12.5mm, 25mm, 50mm)Loads up to 25kg (>25kg possible if required)DC direct drive with single eccentric (no belt)100% stainless steel interiorDimensions: H771 x W1275 x D845mmWeight: 210kgFIVE YEAR WARRANTY HARDWARE1 YEAR WARRANTY ON SENSORS