Learn More
MSC Vikan Lobby Dustpan with Broom, White, 14.2 in, 1 ea
Catalog No. 502819693
Encompass
The Vikan Lobby Dustpan with Broom is designed for efficient and ergonomic floor cleanup in professional environments. Its thoughtful construction minimizes physical strain during use and ensures thorough sanitation.
  • Reduces physical strain with 37 in handle
  • Large 14.2 in x 7 in bin holds optimal amount of dirt
  • Easy to disassemble for washing and sanitizing
  • Built without hard edges to reduce residue and bacteria buildup
  • Ideal for efficient sweeping on dry floors
  • Durable polypropylene, anodized aluminum, and PBT bristle construction
  • Complies with FDA Regulation CFR 21, Halal, and Kosher
Catalog No. 50-281-9693 Supplier MSC Supplier No. 32587339
The Vikan Lobby Dustpan with Broom is designed for efficient and ergonomic floor cleanup in professional environments. Its thoughtful construction minimizes physical strain during use and ensures thorough sanitation.
  • Reduces physical strain with 37 in handle
  • Large 14.2 in x 7 in bin holds optimal amount of dirt
  • Easy to disassemble for washing and sanitizing
  • Built without hard edges to reduce residue and bacteria buildup
  • Ideal for efficient sweeping on dry floors
  • Durable polypropylene, anodized aluminum, and PBT bristle construction
  • Complies with FDA Regulation CFR 21, Halal, and Kosher