Learn More
Medchemexpress LLC Lobeglitazone (sulfate) | 763108-62-9 | 99.6% | 578.61 | 50 MG
SDP
Catalog No. 5000360971
Encompass_Preferred
Lobeglitazone sulfate is a new type of thiazolidinedione and an orally active agonist for PPAR with EC50 of 137.4 nM and 546.3 nM for PPARγ and PPARα. It is an inhibitor for the ERK/JNK/Smad/NF-κB signaling pathway and exhibits anti-inflammatory, anti-diabetic, anti-fibrotic, and anti-atherosclerotic properties.
  • Orally active agonist for PPAR.
  • Inhibits the ERK/JNK/Smad/NF-κB signaling pathway.
  • Exhibits anti-inflammatory properties.
  • Possesses anti-diabetic properties.
  • Demonstrates anti-fibrotic and anti-atherosclerotic properties.
Catalog No. 50-003-60971 Supplier Medchemexpress LLC Supplier No. HY14928A50MG
Lobeglitazone sulfate is a new type of thiazolidinedione and an orally active agonist for PPAR with EC50 of 137.4 nM and 546.3 nM for PPARγ and PPARα. It is an inhibitor for the ERK/JNK/Smad/NF-κB signaling pathway and exhibits anti-inflammatory, anti-diabetic, anti-fibrotic, and anti-atherosclerotic properties.
  • Orally active agonist for PPAR.
  • Inhibits the ERK/JNK/Smad/NF-κB signaling pathway.
  • Exhibits anti-inflammatory properties.
  • Possesses anti-diabetic properties.
  • Demonstrates anti-fibrotic and anti-atherosclerotic properties.