Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lobelanine | 579-21-5 | 5 MG
SDP

Catalog No. 5000417507
Encompass_Preferred

Lobelanine is a chemical precursor that plays a role in the biosynthesis of Lobeline. Lobeline itself is recognized as a partial nicotinic agonist and is utilized as an agent for smoking cessation.

  • For research use only.
  • Available in solid form.
  • Color ranges from white to off-white.
  • Storage at 4°C, protected from light.
  • Stock solution storage at -80°C for 6 months or -20°C for 1 month, protected from light.

Catalog No. 50-004-17507 Supplier Medchemexpress LLC Supplier No. HYN85055MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lobelanine is a chemical precursor that plays a role in the biosynthesis of Lobeline. Lobeline itself is recognized as a partial nicotinic agonist and is utilized as an agent for smoking cessation.

  • For research use only.
  • Available in solid form.
  • Color ranges from white to off-white.
  • Storage at 4°C, protected from light.
  • Stock solution storage at -80°C for 6 months or -20°C for 1 month, protected from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.