Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lobeline (hydrochloride) | 134-63-4 | 100.0% | 500 MG
SDP

Catalog No. 5000409114
Encompass_Preferred

Lobeline (α-Lobeline) hydrochloride is a brain-penetrant nicotinic receptor agonist. It increases dopamine (DA) release by inhibiting DA uptake into synaptic vesicles and altering presynaptic DA storage, making it effective in smoking cessation.

  • Brain-penetrant nicotinic receptor agonist
  • Increases dopamine (DA) release
  • Inhibits DA uptake into synaptic vesicles
  • Alters presynaptic DA storage
  • Effective in smoking cessation
  • Soluble in water at 6 mg/mL
  • White to off-white solid appearance
  • Molecular weight of 373.92
  • Chemical formula C22H28ClNO2
  • Melting/freezing point 183-185 °C

Catalog No. 50-004-09114 Supplier Medchemexpress LLC Supplier No. HYB0979500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lobeline (α-Lobeline) hydrochloride is a brain-penetrant nicotinic receptor agonist. It increases dopamine (DA) release by inhibiting DA uptake into synaptic vesicles and altering presynaptic DA storage, making it effective in smoking cessation.

  • Brain-penetrant nicotinic receptor agonist
  • Increases dopamine (DA) release
  • Inhibits DA uptake into synaptic vesicles
  • Alters presynaptic DA storage
  • Effective in smoking cessation
  • Soluble in water at 6 mg/mL
  • White to off-white solid appearance
  • Molecular weight of 373.92
  • Chemical formula C22H28ClNO2
  • Melting/freezing point 183-185 °C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.