Learn More
Medchemexpress LLC Lobeline (hydrochloride) | 134-63-4 | 99.9% | 50 MG
SDP
Catalog No. 5000408295
Encompass_Preferred
Lobeline hydrochloride is a brain-penetrant nicotinic receptor agonist that increases dopamine release by inhibiting dopamine uptake into synaptic vesicles and altering presynaptic dopamine storage. It is effective in smoking cessation and is intended for research use only.
  • Brain-penetrant nicotinic receptor agonist
  • Increases dopamine release
  • Inhibits dopamine uptake into synaptosomes and vesicles
  • Effective in smoking cessation studies
  • Decreases d-methamphetamine self-administration in rat models
Catalog No. 50-004-08295 Supplier Medchemexpress LLC Supplier No. HYB097950MG
Lobeline hydrochloride is a brain-penetrant nicotinic receptor agonist that increases dopamine release by inhibiting dopamine uptake into synaptic vesicles and altering presynaptic dopamine storage. It is effective in smoking cessation and is intended for research use only.
  • Brain-penetrant nicotinic receptor agonist
  • Increases dopamine release
  • Inhibits dopamine uptake into synaptosomes and vesicles
  • Effective in smoking cessation studies
  • Decreases d-methamphetamine self-administration in rat models