Learn More
Cayman Chemical (–)-Lobelin (hydrochloride), Purity: ≥95%, CAS: 134-63-4, 250 mg
SDP
Catalog No. 502978768
Encompass_Preferred
An alkaloid with diverse biological activities; binds to nAChRs in rat brain homogenates (Ki = 4 nM); has antinociceptive effects in the tail-flick assay in mice; reduces the number of errors in a repeated acquisition procedure in the radial arm maze in rats at 0.3 and 0.9 mg/kg; decreases immobility time in the forced swim test and feeding latency in the novelty suppressed feeding test at 1 and 4 mg/kg
Catalog No. 50-297-8768 Supplier Cayman Chemical Supplier No. 29526250 MG
An alkaloid with diverse biological activities; binds to nAChRs in rat brain homogenates (Ki = 4 nM); has antinociceptive effects in the tail-flick assay in mice; reduces the number of errors in a repeated acquisition procedure in the radial arm maze in rats at 0.3 and 0.9 mg/kg; decreases immobility time in the forced swim test and feeding latency in the novelty suppressed feeding test at 1 and 4 mg/kg