Learn More
Medchemexpress LLC Lobenzarit (disodium) | 64808-48-6 | MFCD00941422 | 99.9% | 335.65 g/mol | C14H8ClNNa2O4 | 50 MG
SDP
Catalog No. 5000241310
Encompass_Preferred
Lobenzarit disodium (CCA) is an immunomodulatory research compound with reported anti-arthritic and antioxidative activity. It is used in preclinical in vitro and in vivo studies to investigate immune modulation and oxidative stress pathways; supplied as the disodium salt for improved solubility and handling. For research use only.
  • Immunomodulator with reported anti-arthritic activity.
  • Antioxidative properties useful in oxidative stress studies.
  • Disodium salt form improves aqueous solubility and handling.
  • Molecular weight 335.65 g/mol; formula C14H8ClNNa2O4.
  • Suitable for preclinical in vitro and in vivo assays; for research use only.
Catalog No. 50-002-41310 Supplier Medchemexpress LLC Supplier No. HY10736050MG
Lobenzarit disodium (CCA) is an immunomodulatory research compound with reported anti-arthritic and antioxidative activity. It is used in preclinical in vitro and in vivo studies to investigate immune modulation and oxidative stress pathways; supplied as the disodium salt for improved solubility and handling. For research use only.
  • Immunomodulator with reported anti-arthritic activity.
  • Antioxidative properties useful in oxidative stress studies.
  • Disodium salt form improves aqueous solubility and handling.
  • Molecular weight 335.65 g/mol; formula C14H8ClNNa2O4.
  • Suitable for preclinical in vitro and in vivo assays; for research use only.