Learn More
Vector Laboratories LOBSTR-BL21(DE3)-RIL, 4x50uL
SDP
Catalog No. NC1789768
Encompass
LOBSTR (low background strain) is an E. coli strain for the expression of recombinant polyhistidine-tagged proteins. This strain has been optimized for one-step downstream polyhistidine-tag affinity purification and is ideal for poorly expressing proteins.
Catalog No. NC1789768 Supplier Vector Laboratories Supplier No. EC1002
LOBSTR (low background strain) is an E. coli strain for the expression of recombinant polyhistidine-tagged proteins. This strain has been optimized for one-step downstream polyhistidine-tag affinity purification and is ideal for poorly expressing proteins.