Learn More
Aviva Systems Biology Corp MYL6B Antibody - N-terminal region (ARP52009_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52009_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC100359980. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: LOC100359980ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Rat
Catalog No. 50-285-624