Learn More
Aviva Systems Biology Corp LOC100360413 Antibody - middle region (ARP48165_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48165_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC100360413. It was validated on Western Blot. . Alternate names: C20orf25, dJ998H6.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-285-348