Learn More
Aviva Systems Biology Corp LOC100364462 ANTIBODY 50UG/50U
SDP
Supplier:  Aviva Systems Biology Corp ARP51951_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC100364462. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human LOC100364462. Alternate names: LOC100364462ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-614