Learn More
Aviva Systems Biology Corp LOC100364937 ANTIBODY 50UG/50U
SDP
Supplier:  Aviva Systems Biology Corp ARP52456_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC100364937. It was validated on Western Blot. . Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-717