Learn More
Aviva Systems Biology Corp LOC115648 Antibody - middle region (ARP36158_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36158_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC115648. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LOC115648. Alternate names: -ApplicationWB. Species reactivity: Human, Mouse
Catalog No. 50-014-56037