Learn More
Aviva Systems Biology Corp ABHD15 Antibody - N-terminal region (ARP53458_P050) | 100uL
Supplier: Aviva Systems Biology Corp ARP53458_P050
This is a rabbit polyclonal antibody against LOC116236. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LOC116236. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat