Learn More
Aviva Systems Biology Corp EXOC3-AS1 Antibody - N-terminal region (ARP52617_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52617_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC116349. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LOC116349. Alternate names: -ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60786