Learn More
Aviva Systems Biology Corp ZNF57 Antibody - N-terminal region (ARP34891_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34891_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF57. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF57. Alternate names: ZNF424ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig
Catalog No. 50-014-55602