Learn More
TARGETMOL CHEMICALS INC LOC14 25MG
SDP
Catalog No. 502579748
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. LOC14 to be the most potent PDI inhibitor reported to date( EC50 500 nM)purity: 98%
Catalog No. 50-257-9748 Supplier TARGETMOL CHEMICALS INC Supplier No. T553425MG
Also available in 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. LOC14 to be the most potent PDI inhibitor reported to date( EC50 500 nM)purity: 98%