Learn More
Aviva Systems Biology Corp MGAT4D Antibody - middle region (ARP56050_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56050_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC152586. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human LOC152586. Alternate names: MGC43429ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse
Catalog No. 50-286-113