Learn More
Aviva Systems Biology Corp FAM153B Antibody - middle region (ARP51873_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51873_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC202134. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LOC202134. Alternate names: DKFZp434D115ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60634