Learn More
Aviva Systems Biology Corp VMA21 Antibody - N-terminal region (ARP44721_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44721_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC203547. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human LOC203547. Alternate names: MEAX, XMEAApplicationIHC, WB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58884