Learn More
Aviva Systems Biology Corp EVX2 Antibody - N-terminal region (ARP31982_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31982_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC344191. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LOC344191. Alternate names: EVX-2ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-54403