Learn More
Aviva Systems Biology Corp ZNF391 Antibody - N-terminal region (ARP31983_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31983_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC346157. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human LOC346157. Alternate names: dJ153G14.3ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Pig
Catalog No. 50-014-52848