Learn More
Aviva Systems Biology Corp C2orf68 Antibody - N-terminal region (ARP54429_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54429_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC388969. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LOC388969. Alternate names: FLJ14112, FLJ35653, MGC131675ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61204