Learn More
Aviva Systems Biology Corp ONECUT3 Antibody - C-terminal region (ARP34502_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34502_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC390874. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human LOC390874. Alternate names: OC3ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat
Catalog No. 50-014-55422