Learn More
Aviva Systems Biology Corp FAM174B Antibody - C-terminal region (ARP42751_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42751_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC400451. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human LOC400451. Alternate names: MGC102891ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Yeast, Zebrafish
Catalog No. 50-014-58328