Learn More
Aviva Systems Biology Corp ZNF833P Antibody - middle region (ARP50380_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50380_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC401898. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LOC401898. Alternate names: ZNF833ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53628