Learn More
Aviva Systems Biology Corp C7orf61 Antibody - C-terminal region (ARP54388_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54388_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC402573. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human LOC402573. Alternate names: -ApplicationWB. Species reactivity: Human
Catalog No. 50-014-61194