Learn More
Aviva Systems Biology Corp LOC632209 ANTIBODY 50UG/50UL
SDP
Supplier:  Aviva Systems Biology Corp ARP44671_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC632209. It was validated on Western Blot. . Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-157