Learn More
Aviva Systems Biology Corp ZNF862 Antibody - N-terminal region (ARP50475_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50475_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC643641. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LOC643641. Alternate names: FLJ30362, KIAA0543ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Rabbit, Rat
Catalog No. 50-014-60352