Learn More
Aviva Systems Biology Corp LOC647589 Antibody - C-terminal region (ARP51075_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51075_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC647589. It was validated on Western Blot. . Alternate names: LOC647589ApplicationWB. Species reactivity: Human
Catalog No. 50-285-555