Learn More
Aviva Systems Biology Corp ZSCAN5C Antibody - C-terminal region (ARP50970_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50970_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC649137. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human LOC649137. Alternate names: ZNF495CApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60477