Learn More
Aviva Systems Biology Corp LOC683991 ANTIBODY 50UG/50UL
SDP
Supplier:  Aviva Systems Biology Corp ARP58537_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC683991. It was validated on Western Blot. . Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-286-481