Learn More
Aviva Systems Biology Corp ATP6AP1L Antibody - C-terminal region (ARP44720_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44720_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC92270. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human LOC92270. Alternate names: MGC138396ApplicationWB. Species reactivity: Dog, Horse, Human, Pig
Catalog No. 50-014-58883