Learn More
Aviva Systems Biology Corp SP140L Antibody - middle region (ARP36135_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36135_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC93349. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LOC93349. Alternate names: ApplicationWB. Species reactivity: Horse, Human, Rat
Catalog No. 50-014-53034