Learn More
Grainger Lock-On Air Chuck
Catalog No. 502750826
Encompass_Preferred
The Haltec Lock-On Air Chuck Lock-On 140X12 is an air chuck that locks onto the air hose to provide a secure connection. It has a 1/4 inch NPT female inlet and a 1/4 inch male outlet. The chuck body and sleeve are made from zinc alloy with a max pressure rating of 140 psi. This air chuck features a rubber sleeve to protect hoses and fittings from wear. It can be used with air hoses up to 3/8 inch OD. The Lock-On design prevents accidental disconnection of air hoses. This is a convenient and durable air chuck for industrial applications.
Catalog No. 50-275-0826 Supplier Grainger Supplier No. 140X12
The Haltec Lock-On Air Chuck Lock-On 140X12 is an air chuck that locks onto the air hose to provide a secure connection. It has a 1/4 inch NPT female inlet and a 1/4 inch male outlet. The chuck body and sleeve are made from zinc alloy with a max pressure rating of 140 psi. This air chuck features a rubber sleeve to protect hoses and fittings from wear. It can be used with air hoses up to 3/8 inch OD. The Lock-On design prevents accidental disconnection of air hoses. This is a convenient and durable air chuck for industrial applications.