Learn More
Grainger Lock-On Air Chuck Steel
Catalog No. 502750692
Encompass_Preferred
The Haltec Lock-On Air Chuck is an air chuck that allows for quick connection and disconnection of air hoses. It has a 1/4" NPT female inlet and features a lock-on coupler that securely grips standard air hose fittings. The chuck body and collar are made from zinc alloy for durability. This air chuck provides a convenient way to quickly attach air hoses for inflating tires or powering air tools. Its lock-on design ensures a tight seal and prevents accidental disconnections.
Catalog No. 50-275-0692 Supplier Grainger Supplier No. 140X11
The Haltec Lock-On Air Chuck is an air chuck that allows for quick connection and disconnection of air hoses. It has a 1/4" NPT female inlet and features a lock-on coupler that securely grips standard air hose fittings. The chuck body and collar are made from zinc alloy for durability. This air chuck provides a convenient way to quickly attach air hoses for inflating tires or powering air tools. Its lock-on design ensures a tight seal and prevents accidental disconnections.