Learn More
Kurin™ Lock for VersaTREK™ REDOX™ 21 Gauge Push Button
SDP
Catalog No. 23232014
Change view
Click to view available options
Content And Storage:
Room Temperature
Catalog No. Content And Storage
23-232-014 Room Temperature
1 options
This item is not returnable. View return policy
Catalog No. 23-232-014 Supplier Kurin™ Supplier No. T21221
Add to Cart
Add to Cart
This item is not returnable. View return policy
Kurin™ blood culture collection set is the simple solution to putting skin contaminants on the sideline.
  • FDA 510(k) cleared
  • Blood conserving for Peds
  • Prepackaged for ease of use
  • No change in workflow
  • Supports antimicrobial stewardship
  • Each Kurin™ blood culture collection set features industry-leading butterfly needles
  • Compatible with all major blood culture bottles
Specifications
Content And Storage Room Temperature