Learn More
Waters Corp Waters Corporation Compression Screw, Locking, Dual Thread, Gold-Plated Stainless Steel
Catalog No. NC2241341 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This compression screw is a locking, dual-thread, two-piece gold-plated stainless steel component designed for use in Active Pre-Heater systems. It is capable of withstanding pressures up to 18,000 psi and replaces existing compression screws, working in conjunction with PEEK ferrules.
  • Two-piece design.
  • Gold-plated stainless steel construction.
  • Replaces existing compression screws on Active Pre-Heaters.
  • Withstands pressures up to 18,000 psi.
  • Compatible with PEEK ferrules.
Catalog No. NC2241341 Supplier Waters Corp Supplier No. 410003354
This compression screw is a locking, dual-thread, two-piece gold-plated stainless steel component designed for use in Active Pre-Heater systems. It is capable of withstanding pressures up to 18,000 psi and replaces existing compression screws, working in conjunction with PEEK ferrules.
  • Two-piece design.
  • Gold-plated stainless steel construction.
  • Replaces existing compression screws on Active Pre-Heaters.
  • Withstands pressures up to 18,000 psi.
  • Compatible with PEEK ferrules.